Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ifnphi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207640.1 | NP_997523.1 | 4 - 5 | 487 | 114 | |
GenBank mRNA | AB093588.1 | - | 5 - 6 | 495 | 114 | |
AJ544822.1 | - | 4 - 5 | 457 | 114 | ||
AY135716.1 | - | 4 - 5 | 457 | 114 | ||
BC162493.1 | - | 4 - 5 | 470 | 114 | ||
EF014960.1 | - | 5 - 6 | 441 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |