Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212673.1 | NP_997838.1 | 4 - 5 | 326 | 124 | |
GenBank mRNA | AY499002.1 | - | 3 - 4 | 266 | 124 | |
BC052130.1 | - | 4 - 5 | 326 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |