Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fahd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212918.1 | NP_998083.1 | 2 - 3 | 380 | 125 | |
XM_021478226.1 | XP_021333901.1 | 2 - 3 | 292 | 125 | ||
XM_021478227.1 | XP_021333902.1 | 2 - 3 | 441 | 125 | ||
XM_021478228.1 | XP_021333903.1 | 2 - 3 | 292 | 125 | ||
XM_021478229.1 | XP_021333904.1 | 2 - 3 | 376 | 125 | ||
GenBank mRNA | BC067576.1 | - | 2 - 3 | 380 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |