Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: caprin1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213068.1 | NP_998233.1 | 6 - 7 | 786 | 93 | |
XM_021467626.1 | XP_021323301.1 | 6 - 7 | 93 | |||
XM_021467627.1 | XP_021323302.1 | 6 - 7 | 93 | |||
GenBank mRNA | BC047196.1 | - | 6 - 7 | 786 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |