Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213319.2 | NP_998484.1 | 6 - 7 | 1074 | 55 | |
GenBank mRNA | BC057409.1 | - | 6 - 7 | 1074 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |