Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxw5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213319.2 | NP_998484.1 | 9 - 10 | 1623 | 124 | |
GenBank mRNA | BC057409.1 | - | 9 - 10 | 1623 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |