Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: adgrl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213367.2 | NP_998532.2 | 9 - 10 | 1245 | 108 | |
GenBank mRNA | BC055171.1 | - | 9 - 10 | 1242 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |