Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cul1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213495.1 | NP_998660.1 | 14 - 15 | 1864 | 93 | |
XM_021470126.1 | XP_021325801.1 | 14 - 15 | 1951 | 93 | ||
GenBank mRNA | BC048370.1 | - | 14 - 15 | 1864 | 93 | |
BC165368.1 | - | 13 - 14 | 1630 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |