Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sirt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002071.1 | NP_001002071.1 | 7 - 8 | 829 | 98 | |
XM_021479673.1 | XP_021335348.1 | 7 - 8 | 846 | 98 | ||
XM_021479674.1 | XP_021335349.1 | 7 - 8 | 691 | 98 | ||
XM_021479677.1 | XP_021335352.1 | 5 - 6 | 571 | 98 | ||
GenBank mRNA | BC071405.1 | - | 7 - 8 | 829 | 98 | |
BC153591.1 | - | 7 - 8 | 796 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |