Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elavl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002172.2 | NP_001002172.2 | 5 - 6 | 698 | 79 | |
GenBank mRNA | BC072716.1 | - | 5 - 6 | 654 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |