Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ltv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002316.2 | NP_001002316.2 | 2 - 3 | 305 | 65 | |
XM_021470934.1 | XP_021326609.1 | 2 - 3 | 300 | 65 | ||
GenBank mRNA | AY648712.1 | - | 2 - 3 | 263 | 65 | |
BC085516.1 | - | 2 - 3 | 278 | 65 | ||
EH504800.1 | - | 2 - 3 | 316 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |