Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rdh12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002325.1 | NP_001002325.1 | 2 - 3 | 217 | 91 | |
GenBank mRNA | BC076473.1 | - | 2 - 3 | 217 | 91 | |
BC163997.1 | - | 2 - 3 | 234 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |