Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002603.2 | NP_001002603.1 | 4 - 5 | 650 | 70 | |
XM_005165699.3 | XP_005165756.1 | 5 - 6 | 708 | 70 | ||
GenBank mRNA | BC075992.1 | - | 4 - 5 | 654 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |