Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: naxe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002618.1 | NP_001002618.1 | 6 - 7 | 823 | 62 | |
GenBank mRNA | BC075969.1 | - | 6 - 7 | 823 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |