Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003425.2 | NP_001003425.2 | 2 - 3 | 193 | 115 | |
GenBank mRNA | BC076243.1 | - | 2 - 3 | 157 | 115 | |
EH505535.1 | - | 2 - 3 | 189 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |