Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lysmd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003507.1 | NP_001003507.1 | 2 - 3 | 786 | 91 | |
GenBank mRNA | BC077165.1 | - | 2 - 3 | 786 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |