Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hibadha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003646.2 | NP_001003646.1 | 3 - 4 | 364 | 85 | |
XM_009294159.3 | XP_009292434.1 | 3 - 4 | 427 | 85 | ||
XM_021468051.1 | XP_021323726.1 | 3 - 4 | 427 | 85 | ||
GenBank mRNA | BC078207.1 | - | 3 - 4 | 367 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |