Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hibadha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003646.2 | NP_001003646.1 | 7 - 8 | 843 | 67 | |
GenBank mRNA | BC078207.1 | - | 7 - 8 | 846 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |