Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbgt1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004620.1 | NP_001004620.1 | 3 - 4 | 268 | 124 | |
XM_021476097.1 | XP_021331772.1 | 3 - 4 | 274 | 124 | ||
GenBank mRNA | BC081406.1 | - | 3 - 4 | 268 | 124 | |
BC164153.1 | - | 3 - 4 | 149 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |