Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gbgt1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004620.1 | NP_001004620.1 | 4 - 5 | 322 | 74 | |
XM_021476095.1 | XP_021331770.1 | 4 - 5 | 435 | 74 | ||
XM_021476097.1 | XP_021331772.1 | 4 - 5 | 328 | 74 | ||
GenBank mRNA | BC081406.1 | - | 4 - 5 | 322 | 74 | |
BC164153.1 | - | 4 - 5 | 203 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |