Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phyhiplb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005602.2 | NP_001005602.2 | 3 - 4 | 696 | 67 | |
XM_005158004.4 | XP_005158061.1 | 2 - 3 | 388 | 67 | ||
XM_005158006.4 | XP_005158063.1 | 2 - 3 | 357 | 67 | ||
XM_017358413.2 | XP_017213902.1 | 2 - 3 | 315 | 67 | ||
XM_021480001.1 | XP_021335676.1 | 2 - 3 | 506 | 67 | ||
GenBank mRNA | BC082295.1 | - | 2 - 3 | 324 | 67 | |
BC139549.1 | - | 3 - 4 | 696 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |