Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: galk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006002.1 | NP_001006002.1 | 7 - 8 | 1309 | 111 | |
GenBank mRNA | BC083195.1 | - | 7 - 8 | 1309 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |