Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mrps34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007376.2 | NP_001007377.2 | 3 - 4 | 443 | 126 | |
GenBank mRNA | BC085462.1 | - | 3 - 4 | 438 | 126 | |
BC153565.1 | - | 2 - 3 | 390 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |