Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: krcp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012366.1 | NP_001012366.1 | 2 - 3 | 277 | 99 | |
XM_005172254.4 | XP_005172311.1 | 2 - 3 | 353 | 99 | ||
GenBank mRNA | AY835383.1 | - | 2 - 3 | 277 | 99 | |
BC108043.1 | - | 2 - 3 | 341 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |