Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rpp30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013479.1 | NP_001013497.1 | 2 - 3 | 219 | 113 | |
XM_021480064.1 | XP_021335739.1 | 2 - 3 | 72 | 113 | ||
GenBank mRNA | BC091449.1 | - | 2 - 3 | 219 | 113 | |
BC164838.1 | - | 2 - 3 | 169 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |