Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: exosc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017585.1 | NP_001017585.1 | 4 - 5 | 450 | 127 | |
GenBank mRNA | BC092997.1 | - | 4 - 5 | 450 | 127 | |
BC152123.1 | - | 4 - 5 | 446 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |