Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnmt3bb.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020479.1 | NP_001018315.1 | 4 - 5 | 1689 | 97 | |
XM_005162039.4 | XP_005162096.1 | 4 - 5 | 1745 | 97 | ||
XM_021469750.1 | XP_021325425.1 | 3 - 4 | 1580 | 97 | ||
GenBank mRNA | AB196916.1 | - | 4 - 5 | 1689 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |