Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ncs1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020514.2 | NP_001018350.1 | 6 - 7 | 1034 | 76 | |
XM_021478256.1 | XP_021333931.1 | 7 - 8 | 1924 | 76 | ||
GenBank mRNA | AY605665.1 | - | 6 - 7 | 489 | 76 | |
BC116500.1 | - | 6 - 7 | 1034 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |