Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctbs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020729.1 | NP_001018565.1 | 2 - 3 | 339 | 81 | |
XM_017358228.2 | XP_017213717.1 | 2 - 3 | 81 | |||
GenBank mRNA | BC072723.1 | - | 2 - 3 | 81 | ||
BC095064.1 | - | 2 - 3 | 339 | 81 | ||
BC164759.1 | - | 2 - 3 | 285 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |