Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2y3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020822.1 | NP_001018658.1 | 4 - 5 | 668 | 107 | |
GenBank mRNA | AY927850.1 | - | 4 - 5 | 664 | 107 | |
BC095374.1 | - | 4 - 5 | 668 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |