Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2y3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020822.1 | NP_001018658.1 | 6 - 7 | 986 | 116 | |
GenBank mRNA | AY927850.1 | - | 6 - 7 | 982 | 116 | |
BC095374.1 | - | 6 - 7 | 986 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |