Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2y3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020822.1 | NP_001018658.1 | 7 - 8 | 1175 | 89 | |
GenBank mRNA | BC095374.1 | - | 7 - 8 | 1175 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |