Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eif3m | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024735.1 | NP_001019906.1 | 5 - 6 | 620 | 80 | |
GenBank mRNA | AY398350.1 | - | 5 - 6 | 620 | 80 | |
BC053188.1 | - | 5 - 6 | 584 | 80 | ||
BC164471.1 | - | 5 - 6 | 564 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |