Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scamp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025525.1 | NP_001020696.1 | 6 - 7 | 622 | 82 | |
GenBank mRNA | BC097198.1 | - | 6 - 7 | 622 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |