Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001043334.1 | NP_001036799.1 | 6 - 7 | 791 | 76 | |
GenBank mRNA | BC163370.1 | - | 6 - 7 | 865 | 76 | |
BC163373.1 | - | 6 - 7 | 866 | 76 | ||
DQ273166.1 | - | 6 - 7 | 791 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |