Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdgfab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076757.1 | NP_001070225.1 | 2 - 3 | 649 | 129 | |
XM_005163736.4 | XP_005163793.1 | 2 - 3 | 700 | 129 | ||
GenBank mRNA | BC124288.1 | - | 2 - 3 | 649 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |