Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gabra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077326.1 | NP_001070794.1 | 6 - 7 | 923 | 74 | |
XM_009295685.3 | XP_009293960.1 | 6 - 7 | 676 | 74 | ||
XM_009295686.3 | XP_009293961.1 | 6 - 7 | 1008 | 74 | ||
XM_009295687.3 | XP_009293962.1 | 6 - 7 | 1158 | 74 | ||
GenBank mRNA | BC124697.1 | - | 6 - 7 | 923 | 74 | |
BC164540.1 | - | 5 - 6 | 596 | 74 | ||
FJ915065.1 | - | 4 - 5 | 464 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |