Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: crfb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079681.1 | NP_001073149.1 | 5 - 6 | 613 | 142 | |
GenBank mRNA | BC163577.1 | - | 5 - 6 | 586 | 142 | |
BC163583.1 | - | 5 - 6 | 586 | 142 | ||
EF014952.1 | - | 5 - 6 | 613 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |