Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp4v7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079996.1 | NP_001073465.1 | 2 - 3 | 380 | 103 | |
GenBank mRNA | BC125940.1 | - | 2 - 3 | 380 | 103 | |
BC165384.1 | - | 2 - 3 | 347 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |