Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rad21l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080050.1 | NP_001073519.1 | 3 - 4 | 341 | 70 | |
XM_021478297.1 | XP_021333972.1 | 2 - 3 | 372 | 70 | ||
GenBank mRNA | BC125933.1 | - | 3 - 4 | 341 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |