Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scxa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083069.2 | NP_001076538.2 | 1 - 2 | 890 | 61 | |
GenBank mRNA | EH579808.1 | - | 1 - 2 | 899 | 61 | |
EH611882.1 | - | 1 - 2 | 595 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Probe spans exons |