Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: col11a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083844.1 | NP_001077313.1 | 20 - 21 | 1984 | 82 | |
XM_005162813.1 | XP_005162870.1 | 21 - 22 | 2311 | 82 | ||
XM_005162814.1 | XP_005162871.1 | 21 - 22 | 2308 | 82 | ||
XM_005162815.1 | XP_005162872.1 | 20 - 21 | 2224 | 82 | ||
XM_005162818.1 | XP_005162875.1 | 20 - 21 | 2014 | 82 | ||
XM_021470209.1 | XP_021325884.1 | 19 - 20 | 1927 | 82 | ||
XM_021470210.1 | XP_021325885.1 | 14 - 15 | 1091 | 82 | ||
GenBank mRNA | BC133083.1 | - | 20 - 21 | 1984 | 82 | |
GQ485664.1 | - | 21 - 22 | 2205 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |