Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spns2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083847.1 | NP_001077316.1 | 9 - 10 | 1196 | 126 | |
GenBank mRNA | AB441164.1 | - | 9 - 10 | 1196 | 126 | |
DQ360111.1 | - | 9 - 10 | 1196 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |