Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankrd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102388.1 | NP_001095858.1 | 3 - 4 | 291 | 115 | |
GenBank mRNA | BC150367.1 | - | 3 - 4 | 291 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |