Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankrd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102388.1 | NP_001095858.1 | 7 - 8 | 701 | 93 | |
GenBank mRNA | BC150367.1 | - | 7 - 8 | 701 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |