Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: siglec15l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001102649.1 | NP_001096119.1 | 1 - 2 | 84 | 106 | |
GenBank mRNA | DQ471942.1 | - | 1 - 2 | 84 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |