Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lhx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131208.1 | NP_571283.1 | 909 | 76 | ||
GenBank mRNA | BC162627.1 | - | 876 | 76 | ||
U34590.1 | - | 909 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio |