Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sox11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131336.1 | NP_571411.1 | 1 | 945 | 102 | |
GenBank mRNA | AJ237812.1 | - | 1 | 945 | 102 | |
BC115164.1 | - | 1 | 971 | 102 | ||
BC139605.1 | - | 1 | 951 | 102 | ||
BC164369.1 | - | 1 | 905 | 102 | ||
U85090.1 | - | 1 | 910 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |