Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bambia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131784.2 | NP_571859.1 | 2 - 3 | 671 | 68 | |
GenBank mRNA | AF239925.1 | - | 2 - 3 | 675 | 68 | |
BC059419.1 | - | 2 - 3 | 664 | 68 | ||
BC071292.1 | - | 2 - 3 | 665 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |