Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dachb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152956.2 | NP_694488.2 | 4 - 5 | 936 | 87 | |
XM_009305776.3 | XP_009304051.1 | 4 - 5 | 967 | 87 | ||
XM_021477542.1 | XP_021333217.1 | 3 - 4 | 349 | 87 | ||
GenBank mRNA | AF427109.1 | - | 4 - 5 | 910 | 87 | |
BC163528.1 | - | 4 - 5 | 903 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |